COM448685975 Combi-Blocks, Inc.

1,4-dimethyl (2S,3S)-2,3-bis[(4-methylphenyl)carbonyloxy]butanedioate

  • MDL number MFCD27578193
  • Cas Number : 1773494-04-4
  • Molecular Weight : 414.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0209

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0209
Availability details for
Quantity
  • Availability
  • Add to Cart