COM448685977 Combi-Blocks, Inc.

methyl 3-hydroxy-2-methylquinoline-4-carboxylate

  • MDL number MFCD27578194
  • Cas Number : 104179-54-6
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0212

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0212
Availability details for
Quantity
  • Availability
  • Add to Cart