COM448685983 Combi-Blocks, Inc.

methyl (2R)-oxolane-2-carboxylate

  • MDL number MFCD27578196
  • Cas Number : 87324-00-3
  • Molecular Weight : 130.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0227
Availability details for
Quantity
  • Availability
  • Add to Cart