COM448685989 Combi-Blocks, Inc.

H-Lys(Boc)OMe

  • MDL number MFCD27578198
  • Cas Number : 3017-32-1
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0248
Availability details for
Quantity
  • Availability
  • Add to Cart