COM448685992 Combi-Blocks, Inc.

2,3-Dimethoxycinnamic acid methyl ester

  • MDL number MFCD22381382
  • Cas Number : 15854-60-1
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0261
Availability details for
Quantity
  • Availability
  • Add to Cart