COM448685997 Combi-Blocks, Inc.

Methyl 2-(4-fluorophenyl)thiazole-4-carboxylate

  • MDL number MFCD17214758
  • Cas Number : 1314419-46-9
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0288

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0288
Availability details for
Quantity
  • Availability
  • Add to Cart