COM448685998 Combi-Blocks, Inc.

methyl (2R)-2-(hex-5-en-1-yloxy)propanoate

  • MDL number MFCD27578200
  • Cas Number : 1218998-89-0
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0294

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0294
Availability details for
Quantity
  • Availability
  • Add to Cart