COM448686000 Combi-Blocks, Inc.

methyl (2S)-2-(1,3-dioxoisoindol-2-yl)-3-methylbutanoate

  • MDL number MFCD27578201
  • Cas Number : 124729-87-9
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0310
Availability details for
Quantity
  • Availability
  • Add to Cart