COM448686005 Combi-Blocks, Inc.

methyl 2-(pyrrolidin-1-yl)pyridine-4-carboxylate

  • MDL number MFCD27578203
  • Cas Number : 1692717-97-7
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0325
Availability details for
Quantity
  • Availability
  • Add to Cart