COM448686008 Combi-Blocks, Inc.

methyl 2-(5-bromo-4-chloro-2-nitrophenyl)acetate

  • MDL number MFCD27578204
  • Cas Number : 1820703-56-7
  • Molecular Weight : 308.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0333
Availability details for
Quantity
  • Availability
  • Add to Cart