COM448686010 Combi-Blocks, Inc.

methyl (1R,4R)-bicyclo[2.2.1]hept-5-ene-2-carboxylate

  • MDL number MFCD27578205
  • Cas Number : 1820598-65-9
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0339

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0339
Availability details for
Quantity
  • Availability
  • Add to Cart