COM448686018 Combi-Blocks, Inc.

Methyl 7-methylimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD16875908
  • Cas Number : 1220397-16-9
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0376
Availability details for
Quantity
  • Availability
  • Add to Cart