COM448686023 Combi-Blocks, Inc.

Methyl 6-bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD22422199
  • Cas Number : 1820641-83-5
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0400
Availability details for
Quantity
  • Availability
  • Add to Cart