COM448686024 Combi-Blocks, Inc.

3-Hydroxy-L-Tyrosine methyl ester

  • MDL number MFCD00870911
  • Cas Number : 7101-51-1
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0401

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0401
Availability details for
Quantity
  • Availability
  • Add to Cart