COM448686027 Combi-Blocks, Inc.

Methyl 3-ethanesulfonamidobenzoate

  • MDL number MFCD05116679
  • Cas Number : 834869-20-4
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0410
Availability details for
Quantity
  • Availability
  • Add to Cart