COM448686029 Combi-Blocks, Inc.

Methyl (2s)-2-[(methoxycarbonyl)amino]-3,3-dimethylbutanoate

  • MDL number MFCD23105771
  • Cas Number : 1820570-23-7
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0414
Availability details for
Quantity
  • Availability
  • Add to Cart