COM448686032 Combi-Blocks, Inc.

3-tert-butyl 4-methyl (4R)-1,3-thiazolidine-3,4-dicarboxylate

  • MDL number MFCD27578207
  • Cas Number : 160436-61-3
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0418
Availability details for
Quantity
  • Availability
  • Add to Cart