COM448686034 Combi-Blocks, Inc.

3,5-Bis(trifluoromethyl)phenyl acetic acid methyl ester

  • MDL number MFCD12025301
  • Cas Number : 95299-16-4
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0425
Availability details for
Quantity
  • Availability
  • Add to Cart