COM448686035 Combi-Blocks, Inc.

Methyl 2-(benzyloxycarbonylamino)-2-methylpropanoate

  • MDL number MFCD23378842
  • Cas Number : 172992-10-8
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0426
Availability details for
Quantity
  • Availability
  • Add to Cart