COM448686037 Combi-Blocks, Inc.

methyl 5-chloro-2-acetamidopyridine-4-carboxylate

  • MDL number MFCD27578208
  • Cas Number : 1820648-39-2
  • Molecular Weight : 228.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0430
Availability details for
Quantity
  • Availability
  • Add to Cart