COM448686038 Combi-Blocks, Inc.

methyl (2S)-2,6-di{[(benzyloxy)carbonyl]amino}hexanoate

  • MDL number MFCD00046779
  • Cas Number : 55592-84-2
  • Molecular Weight : 428.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0433
Availability details for
Quantity
  • Availability
  • Add to Cart