COM448686043 Combi-Blocks, Inc.

methyl (2S,3R)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoate

  • MDL number MFCD27578209
  • Cas Number : 1820574-69-3
  • Molecular Weight : 411.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0446
Availability details for
Quantity
  • Availability
  • Add to Cart