COM448686045 Combi-Blocks, Inc.

5-tert-butyl 1-methyl (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanedioate

  • MDL number MFCD27578211
  • Cas Number : 1820576-16-6
  • Molecular Weight : 439.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0451
Availability details for
Quantity
  • Availability
  • Add to Cart