COM448686046 Combi-Blocks, Inc.

Methyl 8-bromooctanoate

  • MDL number MFCD02258673
  • Cas Number : 26825-92-3
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0456

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0456
Availability details for
Quantity
  • Availability
  • Add to Cart