COM448686051 Combi-Blocks, Inc.

4'-Trifluoromethyl-biphenyl-2-carboxylic acid methyl ester

  • MDL number MFCD04972930
  • Cas Number : 91748-18-4
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0468
Availability details for
Quantity
  • Availability
  • Add to Cart