COM448686052 Combi-Blocks, Inc.

1-benzyl 5-methyl (2R)-2-{[(benzyloxy)carbonyl]amino}pentanedioate

  • MDL number MFCD27578214
  • Cas Number : 1820570-17-9
  • Molecular Weight : 385.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0470
Availability details for
Quantity
  • Availability
  • Add to Cart