COM448686053 Combi-Blocks, Inc.

(2R,4R)-Methyl 4-hydroxypyrrolidine-2-carboxylate

  • MDL number MFCD00797544
  • Cas Number : 114676-47-0
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0472
Availability details for
Quantity
  • Availability
  • Add to Cart