COM448686054 Combi-Blocks, Inc.

methyl 2-(adamantan-1-yl)acetate

  • MDL number MFCD01073581
  • Cas Number : 27174-71-6
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0473
Availability details for
Quantity
  • Availability
  • Add to Cart