COM448686055 Combi-Blocks, Inc.

(R)-1-Benzyl 5-methyl 2-((tert-butoxycarbonyl)amino)pentanedioate

  • MDL number MFCD13184782
  • Cas Number : 883985-06-6
  • Molecular Weight : 351.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0474
Availability details for
Quantity
  • Availability
  • Add to Cart