COM448686057 Combi-Blocks, Inc.

1-methyl 4-prop-2-en-1-yl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578215
  • Cas Number : 170943-09-6
  • Molecular Weight : 409.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0477
Availability details for
Quantity
  • Availability
  • Add to Cart