COM448686061 Combi-Blocks, Inc.

2-tert-butyl 3-methyl (3S)-1H,3H,4H,9H-pyrido[3,4-b]indole-2,3-dicarboxylate

  • MDL number MFCD27578217
  • Cas Number : 1240613-17-5
  • Molecular Weight : 330.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0483
Availability details for
Quantity
  • Availability
  • Add to Cart