COM448686062 Combi-Blocks, Inc.

methyl (2S,3S)-2-(1,3-dioxoisoindol-2-yl)-3-methylpentanoate

  • MDL number MFCD27578218
  • Cas Number : 137649-35-5
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0484
Availability details for
Quantity
  • Availability
  • Add to Cart