COM448686064 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-1-yl)propanoate

  • MDL number MFCD27578219
  • Cas Number : 216753-83-2
  • Molecular Weight : 329.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0487
Availability details for
Quantity
  • Availability
  • Add to Cart