COM448686067 Combi-Blocks, Inc.

methyl (2S)-2,6-bis({[(9H-fluoren-9-ylmethoxy)carbonyl]amino})hexanoate

  • MDL number MFCD27578222
  • Cas Number : 1820574-31-9
  • Molecular Weight : 604.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0490
Availability details for
Quantity
  • Availability
  • Add to Cart