COM448686069 Combi-Blocks, Inc.

Methyl 4-[(9h-fluoren-9-ylmethoxycarbonylamino)methyl]benzoate

  • MDL number MFCD03844660
  • Cas Number : 885266-54-6
  • Molecular Weight : 387.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0493
Availability details for
Quantity
  • Availability
  • Add to Cart