COM448686070 Combi-Blocks, Inc.

Methyl 1-([(tert-butoxy)carbonyl]amino)cyclohexane-1-carboxylate

  • MDL number MFCD24392033
  • Cas Number : 1144505-90-7
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0494
Availability details for
Quantity
  • Availability
  • Add to Cart