COM448686073 Combi-Blocks, Inc.

methyl (2S)-2-{[(benzyloxy)carbonyl]amino}-5-(1,3-dioxoisoindol-2-yl)pentanoate

  • MDL number MFCD27578225
  • Cas Number : 142165-70-6
  • Molecular Weight : 410.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0500
Availability details for
Quantity
  • Availability
  • Add to Cart