COM448686076 Combi-Blocks, Inc.

5-tert-butyl 1-methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate

  • MDL number MFCD27578227
  • Cas Number : 156143-34-9
  • Molecular Weight : 317.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0505

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0505
Availability details for
Quantity
  • Availability
  • Add to Cart