COM448686077 Combi-Blocks, Inc.

methyl (2S)-5-amino-2-{[(benzyloxy)carbonyl]amino}pentanoate

  • MDL number MFCD27578228
  • Cas Number : 137559-30-9
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0507

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0507
Availability details for
Quantity
  • Availability
  • Add to Cart