COM448686078 Combi-Blocks, Inc.

methyl 1-(2-bromophenyl)-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD27578229
  • Cas Number : 1820686-31-4
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0508
Availability details for
Quantity
  • Availability
  • Add to Cart