COM448686079 Combi-Blocks, Inc.

Methyl 4-(benzyloxycarbonylamino)butanoate

  • MDL number MFCD00708599
  • Cas Number : 67706-63-2
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0509
Availability details for
Quantity
  • Availability
  • Add to Cart