COM448686081 Combi-Blocks, Inc.

methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(methylsulfanyl)propanoate

  • MDL number MFCD27578230
  • Cas Number : 82835-08-3
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0512

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0512
Availability details for
Quantity
  • Availability
  • Add to Cart