COM448686082 Combi-Blocks, Inc.

1,5-dimethyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanedioate

  • MDL number MFCD27578231
  • Cas Number : 1820572-04-0
  • Molecular Weight : 397.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0513
Availability details for
Quantity
  • Availability
  • Add to Cart