COM448686083 Combi-Blocks, Inc.

methyl (2S)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate

  • MDL number MFCD27578232
  • Cas Number : 951324-28-0
  • Molecular Weight : 397.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0514
Availability details for
Quantity
  • Availability
  • Add to Cart