COM448686084 Combi-Blocks, Inc.

Methyl 2,4,5-trimethoxybenzoate

  • MDL number MFCD00086328
  • Cas Number : 20029-76-9
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0515
Availability details for
Quantity
  • Availability
  • Add to Cart