COM448686085 Combi-Blocks, Inc.

1-benzyl 5-methyl (2S)-2-{[(benzyloxy)carbonyl]amino}pentanedioate

  • MDL number MFCD27578233
  • Cas Number : 105299-31-8
  • Molecular Weight : 385.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0516

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0516
Availability details for
Quantity
  • Availability
  • Add to Cart