COM448686087 Combi-Blocks, Inc.

methyl 2,5-diaminopentanoate hydrochloride

  • MDL number MFCD27578235
  • Cas Number : 261522-14-9
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0519
Availability details for
Quantity
  • Availability
  • Add to Cart