COM448686090 Combi-Blocks, Inc.

methyl (2R)-3-cyclohexyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate

  • MDL number MFCD27578237
  • Cas Number : 1820583-21-8
  • Molecular Weight : 407.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0523
Availability details for
Quantity
  • Availability
  • Add to Cart