COM448686091 Combi-Blocks, Inc.

methyl (2S)-3-cyclohexyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate

  • MDL number MFCD27578238
  • Cas Number : 1500080-83-0
  • Molecular Weight : 407.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0524
Availability details for
Quantity
  • Availability
  • Add to Cart