COM448686092 Combi-Blocks, Inc.

(R)-Methyl 2-(benzyloxycarbonylamino)-2-phenylacetate

  • MDL number MFCD13190420
  • Cas Number : 147780-60-7
  • Molecular Weight : 299.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0525

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0525
Availability details for
Quantity
  • Availability
  • Add to Cart